2,4-Dichloro-6-methylpyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS5187G1
AS5187G5
ACTIAS5187G1EA
0
GBP
ACTIAS5187G1
ACTIAS5187G5
2,4-Dichloro-6-methylpyridine
2,4-Dichloro-6-methylpyridine
Formula:
C₆H₅Cl₂N MW: 162.01 g/mol |
MDL Number:
MFCD08669834 CAS Number: 42779-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...