Methyl-3-fluoropicolinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoro-pyridine-2-carboxylic acid methyl ester, Methyl 3-fluoropyridine-2-carboxylate

AS24284G1 AS24284G5
ACTIAS24284G1EA 0 GBP
ACTIAS24284G1 ACTIAS24284G5
Methyl-3-fluoropicolinate
Methyl 3-fluoropicolinate
Formula: C₇H₆FNO₂
MDL Number: MFCD06659496
CAS Number: 869108-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR