Methyl-3-aminopicolinate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21948G1
AS21948G25
AS21948G5
ACTIAS21948G1EA
0
GBP
ACTIAS21948G1
ACTIAS21948G25
ACTIAS21948G5
Methyl-3-aminopicolinate
Methyl 3-aminopicolinate
Formula:
C₇H₈N₂O₂ |
MDL Number:
MFCD08695602 CAS Number: 36052-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...