Ethyl-3-aminopicolinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21947G1 AS21947G25 AS21947G5
ACTIAS21947G1EA 0 GBP
ACTIAS21947G1 ACTIAS21947G25 ACTIAS21947G5
Ethyl-3-aminopicolinate
Ethyl 3-aminopicolinate
Formula: C₈H₁₀N₂O₂
MDL Number: MFCD00661294
CAS Number: 27507-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR