Methyl-5-chloropicolinate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS33693G1
AS33693G5
ACTIAS33693G1EA
0
GBP
ACTIAS33693G1
ACTIAS33693G5
Methyl-5-chloropicolinate
Methyl 5-chloropicolinate
Formula:
C₇H₆ClNO₂ |
MDL Number:
MFCD09702466 CAS Number: 132308-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...