Ethyl-6-bromopicolinate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

020124-5G 20124-25G 020124-25G
FLUO020124-5GEA 0 GBP
FLUO020124-5G FLUO20124-25G FLUO020124-25G
Ethyl-6-bromopicolinate
Ethyl 6-bromopicolinate
Formula: C₈H₈BrNO₂
MDL Number: MFCD04116913
CAS Number: 21190-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR