Methyl-3-bromopicolinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-bromo-2-pyridinecarboxylate

AS22006G1 AS22006G25 AS22006G5
ACTIAS22006G1EA 14 GBP
ACTIAS22006G1 ACTIAS22006G25 ACTIAS22006G5
Methyl-3-bromopicolinate
Methyl 3-bromopicolinate
Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
MDL Number: MFCD12025946
CAS Number: 53636-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR