5-Isobutyl-1,3-cyclohexanedione

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011173-10G 011173-5G
FLUO011173-10GEA 0 GBP
FLUO011173-10G FLUO011173-5G
5-Isobutyl-1,3-cyclohexanedione
5-Isobutyl-1,3-cyclohexanedione
Formula: C₁₀H₁₆O₂
MDL Number: MFCD00129458
CAS Number: 57641-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR