1-(3-Cyanophenyl)-4-methylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20552G1 AS20552G25 AS20552G5
ACTIAS20552G1EA 0 GBP
ACTIAS20552G1 ACTIAS20552G25 ACTIAS20552G5
1-(3-Cyanophenyl)-4-methylpiperazine
1-(3-Cyanophenyl)-4-methylpiperazine
Formula: C₁₂H₁₅N₃
MDL Number: MFCD07368541
CAS Number: 204078-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR