3-Bromo-5-(trifluoromethoxy)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-5332-5G
BB-5332-1G
COBBBB-5332-5GEA
0
GBP
COBBBB-5332-5G
COBBBB-5332-1G
3-Bromo-5-(trifluoromethoxy)phenylboronic acid
3-Bromo-5-(trifluoromethoxy)phenylboronic acid
Formula:
C₇H₅BBrF₃O₃ |
MDL Number:
MFCD11504853 CAS Number: 1072951-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...