4-Chloro-3-picoline hydrochloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010406-1G 010406-5G
FLUO010406-1GEA 0 GBP
FLUO010406-1G FLUO010406-5G
4-Chloro-3-picoline hydrochloride
4-Chloro-3-picoline hydrochloride
Formula: C₆H₇Cl₂N
MDL Number: MFCD03092890
CAS Number: 19524-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR