4-Chloro-3-picoline hydrochloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS32211G1 AS32211G5
ACTIAS32211G1EA 18 GBP
ACTIAS32211G1 ACTIAS32211G5
4-Chloro-3-picoline hydrochloride
4-Chloro-3-picoline hydrochloride
Formula: C₆H₇Cl₂N
MDL Number: MFCD03092890
CAS Number: 19524-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR