3-(2,2,2-Trifluoroethoxy)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(2,2,2-Trifluoroethoxy)benzeneboronic acid, (3-(2,2,2-Trifluoroethoxy)phenyl)boronic acid

BB-2785-5G BB-2785-1G
COBBBB-2785-5GEA 0 GBP
COBBBB-2785-5G COBBBB-2785-1G
3-(2,2,2-Trifluoroethoxy)phenylboronic acid
3-(2,2,2-Trifluoroethoxy)phenylboronic acid
Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
MDL Number: MFCD07363846
CAS Number: 850593-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR