2-Amino-4,5-dichloro-3-nitropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,5-Dichloro-3-nitropyridin-2-amine

AS12457G1 AS12457G5
ACTIAS12457G1EA 0 GBP
ACTIAS12457G1 ACTIAS12457G5
2-Amino-4,5-dichloro-3-nitropyridine
2-Amino-4,5-dichloro-3-nitropyridine
Formula: C₅H₃Cl₂N₃O₂
MW: 208.0022 g/mol
MDL Number: MFCD15143363
CAS Number: 662116-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR