2-Ethylindene ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B23104.03 B23104.06
B23104.03EA 71.9 GBP
B23104.03 B23104.06
2-Ethylindene ≥97%
2-Ethylindene

Purity: 97% CAS No.: 17059-50-6 Molecular Formula: C11H12 Formula Weight: 144.22 Flash Point: 87°(190°F) Refractive Index: 1.5590 Boiling Point: 94-96°/10mm MDL No.: MFCD01321239

Formula: C₁₁H₁₂
Boiling Pt: 94…96 °C (10 mmHg)
Density: 0.964
Flash Pt: 87 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD01321239
CAS Number: 17059-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR