5-Bromo-2-chloro-4-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51698G1 AS51698G5
ACTIAS51698G1EA 0 GBP
ACTIAS51698G1 ACTIAS51698G5
5-Bromo-2-chloro-4-(tributylstannyl)pyridine
5-Bromo-2-chloro-4-(tributylstannyl)pyridine
Formula: C₁₇H₂₉BrClNSn
MDL Number: MFCD17015005
CAS Number: 821773-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR