4-(4-Cyanophenyl)morpholine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Morpholinobenzonitrile, 4-(4-Morpholinyl)benzonitrile
AS22621G25
AS22621G5
ACTIAS22621G25EA
138
GBP
ACTIAS22621G25
ACTIAS22621G5
4-(4-Cyanophenyl)morpholine
4-(4-Cyanophenyl)morpholine
Formula:
C₁₁H₁₂N₂O MW: 188.23 g/mol |
MDL Number:
MFCD00100115 CAS Number: 10282-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...