4-(2-Cyanophenyl)morpholine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Morpholinobenzonitrile, 2-(Morpholin-4-yl)benzonitrile

AS21306G1 AS21306G25 AS21306G5
ACTIAS21306G1EA 0 GBP
ACTIAS21306G1 ACTIAS21306G25 ACTIAS21306G5
4-(2-Cyanophenyl)morpholine
4-(2-Cyanophenyl)morpholine
Formula: C₁₁H₁₂N₂O
MW: 188.2258 g/mol
MDL Number: MFCD01313390
CAS Number: 204078-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR