Methyl-3-methylthiophene-2-carboxylate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 3-methylthiophene-2-carboxylate
AS21326G25
AS21326G5
ACTIAS21326G25EA
0
GBP
ACTIAS21326G25
ACTIAS21326G5
Methyl-3-methylthiophene-2-carboxylate
Methyl 3-methylthiophene-2-carboxylate
Formula:
C₇H₈O₂S MW: 156.20 g/mol |
MDL Number:
MFCD00234332 CAS Number: 81452-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...