2-Mercaptothiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018767-25G 018767-5G
FLUO018767-25GEA 0 GBP
FLUO018767-25G FLUO018767-5G
2-Mercaptothiophene
2-Mercaptothiophene
Formula: C₄H₄S₂
MDL Number: MFCD00051666
CAS Number: 7774-74-5
EINECS: 231-881-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR