2-Methoxy-6-methylpyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7527-5G
OS-7527-25G
OS-7527-100G
COBBOS-7527-5GEA
0
GBP
COBBOS-7527-5G
COBBOS-7527-25G
COBBOS-7527-100G
2-Methoxy-6-methylpyridine
2-Methoxy-6-methylpyridine
Formula:
C₇H₉NO MW: 123.16 g/mol |
MDL Number:
MFCD04112507 CAS Number: 63071-03-4 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...