2-Formyl-5-picoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Methylpyridine-2-carbaldehyde, 5-Methylpicolinaldehyde

AS50450G1 AS50450G5
ACTIAS50450G1EA 0 GBP
ACTIAS50450G1 ACTIAS50450G5
2-Formyl-5-picoline
2-Formyl-5-picoline
Formula: C₇H₇NO
MW: 121.1400 g/mol
MDL Number: MFCD07367942
CAS Number: 4985-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR