6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, tert-Butyl methyl(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate
PN-8775-1G
PN-8775-250M
COBBPN-8775-1GEA
0
GBP
COBBPN-8775-1G
COBBPN-8775-250M
6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester
6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester
Formula:
C₁₇H₂₇BN₂O₄ MW: 334.22 g/mol |
MDL Number:
MFCD09037484 CAS Number: 1032758-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...