1-Naphthalenemethanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22662G100 AS22662G25
ACTIAS22662G100EA 0 GBP
ACTIAS22662G100 ACTIAS22662G25
1-Naphthalenemethanol
1-Naphthalenemethanol
Formula: C₁₁H₁₀O
MDL Number: MFCD00004044
CAS Number: 4780-79-4
EINECS: 225-324-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR