3-Amino-2-naphthalenemethanol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1359-5G AN-1359-1G
COBBAN-1359-5GEA 0 GBP
COBBAN-1359-5G COBBAN-1359-1G
3-Amino-2-naphthalenemethanol
3-Amino-2-naphthalenemethanol
Formula: C₁₁H₁₁NO
MDL Number: MFCD00155332
CAS Number: 141281-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR