1-(Aminomethyl)-4-bromonaphthalene hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Bromonaphthalen-1-yl)methanamine hydrochloride

AN-1459-5G AN-1459-1G
COBBAN-1459-5GEA 0 GBP
COBBAN-1459-5G COBBAN-1459-1G
1-(Aminomethyl)-4-bromonaphthalene hydrochloride
1-(Aminomethyl)-4-bromonaphthalene hydrochloride
Formula: C₁₁H₁₁BrClN
MW: 272.5687 g/mol
MDL Number: MFCD08706223
CAS Number: 578029-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR