4-tert-Butylcyclohexene-1-ylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2537-1G BB-2537-250M
COBBBB-2537-1GEA 0 GBP
COBBBB-2537-1G COBBBB-2537-250M
4-tert-Butylcyclohexene-1-ylboronic acid
4-tert-Butylcyclohexene-1-ylboronic acid
Formula: C₁₀H₁₉BO₂
MDL Number: MFCD06659856
CAS Number: 850567-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR