4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF1185.1 006632-1G 006632-5G
CHMPCFF1185.1EA 0 GBP
CHMPCFF1185.1 FLUO006632-1G FLUO006632-5G
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid
Formula: C₁₁H₆F₃NO₃
MDL Number: MFCD00236691
CAS Number: 49713-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR