4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20946G25 AS20946G5
ACTIAS20946G25EA 445 GBP
ACTIAS20946G25 ACTIAS20946G5
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid
Formula: C₁₁H₆F₃NO₃
MDL Number: MFCD00236691
CAS Number: 49713-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR