Methyl-6-acetamido-4-chloroquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2512-5G HC-2512-1G
COBBHC-2512-5GEA 0 GBP
COBBHC-2512-5G COBBHC-2512-1G
Methyl-6-acetamido-4-chloroquinoline-2-carboxylate
Methyl 6-acetamido-4-chloroquinoline-2-carboxylate
Formula: C₁₃H₁₁ClN₂O₃
MDL Number: MFCD11855917
CAS Number: 1133115-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR