Methyl-8-bromo-4-hydroxy-5-(trifluoromethyl)quinoline-2-carboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6869-5G
HC-6869-1G
COBBHC-6869-5GEA
0
GBP
COBBHC-6869-5G
COBBHC-6869-1G
Methyl-8-bromo-4-hydroxy-5-(trifluoromethyl)quinoline-2-carboxylate
Methyl 8-bromo-4-hydroxy-5-(trifluoromethyl)quinoline-2-carboxylate
Formula:
C₁₂H₇BrF₃NO₃ |
MDL Number:
MFCD11504926 CAS Number: 1072944-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...