Ethyl-4-hydroxyquinoline-3-carboxylate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Hydroxyquinoline-3-carboxylic acid ethyl ester, Ethyl 4-hydroxyquinoline-3-carboxylate
AS20756G1
AS20756G25
AS20756G5
ACTIAS20756G1EA
0
GBP
ACTIAS20756G1
ACTIAS20756G25
ACTIAS20756G5
Ethyl-4-hydroxyquinoline-3-carboxylate
Ethyl 4-hydroxyquinoline-3-carboxylate
Formula:
C₁₂H₁₁NO₃ MW: 217.2206 g/mol |
MDL Number:
MFCD00173406 CAS Number: 26892-90-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...