Methyl-4-hydroxyquinoline-2-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-hydroxyquinoline-2-carboxylate

AS21047G1 AS21047G5
ACTIAS21047G1EA 0 GBP
ACTIAS21047G1 ACTIAS21047G5
Methyl-4-hydroxyquinoline-2-carboxylate
Methyl 4-hydroxyquinoline-2-carboxylate
Formula: C₁₁H₉NO₃
MW: 203.1900 g/mol
MDL Number: MFCD08693316
CAS Number: 5965-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR