Methyl quinoline-6-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20660G25 AS20660G5
ACTIAS20660G25EA 0 GBP
ACTIAS20660G25 ACTIAS20660G5
Methyl quinoline-6-carboxylate
Methyl quinoline-6-carboxylate
Formula: C₁₁H₉NO₂
MDL Number: MFCD00956766
CAS Number: 38896-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR