Ethyl-7-bromo-4-hydroxyquinoline-3-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6348-5G HC-6348-1G
COBBHC-6348-5GEA 0 GBP
COBBHC-6348-5G COBBHC-6348-1G
Ethyl-7-bromo-4-hydroxyquinoline-3-carboxylate
Ethyl 7-bromo-4-hydroxyquinoline-3-carboxylate
Formula: C₁₂H₁₀BrNO₃
MDL Number: MFCD00487280
CAS Number: 179943-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR