2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

FL10790.2 010790-5G 010790-1G
CHMPFL10790.2EA 0 GBP
CHMPFL10790.2 FLUO010790-5G FLUO010790-1G
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
3,5-Dimethyl-4-hydroxyphenylboronic acid pinacol ester
Formula: C₁₄H₂₁BO₃
MDL Number: MFCD02093724
CAS Number: 269410-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR