Methyl-3,5-dinitro-2-methylbenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20897G25 AS20897G5
ACTIAS20897G25EA 0 GBP
ACTIAS20897G25 ACTIAS20897G5
Methyl-3,5-dinitro-2-methylbenzoate
Methyl 3,5-dinitro-2-methylbenzoate
Formula: C₉H₈N₂O₆
MDL Number: MFCD00229501
CAS Number: 52090-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR