2-[(1',2',3',4'-Tetrahydro-5-naphthyloxy)methyl]phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-8066-1G
BB-8066-250M
COBBBB-8066-1GEA
0
GBP
COBBBB-8066-1G
COBBBB-8066-250M
2-[(1',2',3',4'-Tetrahydro-5-naphthyloxy)methyl]phenylboronic acid
2-[(1',2',3',4'-Tetrahydro-5-naphthyloxy)methyl]phenylboronic acid
Formula:
C₁₇H₁₉BO₃ |
MDL Number:
MFCD07369757 CAS Number: 849062-10-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...