4-Acetyl-3-nitrophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3265-1G BB-3265-250M
COBBBB-3265-1GEA 0 GBP
COBBBB-3265-1G COBBBB-3265-250M
4-Acetyl-3-nitrophenylboronic acid
4-Acetyl-3-nitrophenylboronic acid
Formula: C₈H₈BNO₅
MDL Number: MFCD16618928
CAS Number: 1256345-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR