3-Methoxy-5-(trifluoromethoxy)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-6331-5G BB-6331-1G BB-6331-250M
COBBBB-6331-5GEA 0 GBP
COBBBB-6331-5G COBBBB-6331-1G COBBBB-6331-250M
3-Methoxy-5-(trifluoromethoxy)phenylboronic acid
3-Methoxy-5-(trifluoromethoxy)phenylboronic acid
Formula: C₈H₈BF₃O₄
MDL Number: MFCD12026733
CAS Number: 1256345-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR