3-Butoxy-5-(trifluoromethoxy)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-6361-5G BB-6361-1G
COBBBB-6361-5GEA 0 GBP
COBBBB-6361-5G COBBBB-6361-1G
3-Butoxy-5-(trifluoromethoxy)phenylboronic acid
3-Butoxy-5-(trifluoromethoxy)phenylboronic acid
Formula: C₁₁H₁₄BF₃O₄
MDL Number: MFCD16036112
CAS Number: 1256345-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR