(4-Chlorophenylthio)acetone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-((4-Chlorophenyl)thio)propan-2-one, 1-(4-Chlorophenylthio)propan-2-one

023249-100G 023249-25G
FLUO023249-100GEA 0 GBP
FLUO023249-100G FLUO023249-25G
(4-Chlorophenylthio)acetone
(4-Chlorophenylthio)acetone
Formula: C₉H₉ClOS
MDL Number: MFCD00013644
CAS Number: 25784-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR