3-(Cyclohexylcarbamoyl)-5-nitrophenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-2833-5G
BB-2833-1G
COBBBB-2833-5GEA
0
GBP
COBBBB-2833-5G
COBBBB-2833-1G
3-(Cyclohexylcarbamoyl)-5-nitrophenylboronic acid
3-(Cyclohexylcarbamoyl)-5-nitrophenylboronic acid
Formula:
C₁₃H₁₇BN₂O₅ |
MDL Number:
MFCD07783877 CAS Number: 871332-85-3 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...