1-(Cbz-amino)-4-bromonaphthalene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3174-5G HC-3174-1G
COBBHC-3174-5GEA 0 GBP
COBBHC-3174-5G COBBHC-3174-1G
1-(Cbz-amino)-4-bromonaphthalene
1-(Cbz-Amino)-4-bromonaphthalene
Formula: C₁₈H₁₄BrNO₂
MDL Number: MFCD13195700
CAS Number: 1215206-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR