3-(Methylthio)-1H-1,2,4-triazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009887-1G 009887-5G
FLUO009887-1GEA 0 GBP
FLUO009887-1G FLUO009887-5G
3-(Methylthio)-1H-1,2,4-triazole
3-(Methylthio)-1H-1,2,4-triazole
Formula: C₃H₅N₃S
MDL Number: MFCD01312671
CAS Number: 7411-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR