2-Furoylacetonitrile ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-(Furan-2-yl)-3-oxopropanenitrile, 3-(Fur-2-yl)-3-oxopropanenitrile
A14497.03
A14497.06
A14497.14
A14497.03EA
19.8
GBP
A14497.03
A14497.06
A14497.14
2-Furoylacetonitrile ≥97%
2-Furoylacetonitrile
Purity: 97%
CAS No.: 31909-58-7
Molecular Formula: C7H5NO2
Formula Weight: 135.12
Melting Point: 77-80°
Beilstein Reference: 111902
Formula:
C₇H₅NO₂ MW: 135.12 g/mol Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052210 CAS Number: 31909-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...