5-Bromo-(2,3-methylenedioxy)phenylboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(6-Bromobenzo[d][1,3]dioxol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-5462-5G
PN-5462-1G
COBBPN-5462-5GEA
0
GBP
COBBPN-5462-5G
COBBPN-5462-1G
5-Bromo-(2,3-methylenedioxy)phenylboronic acid pinacol ester
5-Bromo-(2,3-methylenedioxy)phenylboronic acid pinacol ester
Formula:
C₁₃H₁₆BBrO₄ |
MDL Number:
MFCD11855972 CAS Number: 1150271-54-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...