1-(4-Cyanophenyl)piperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21228G25 AS21228G5
ACTIAS21228G25EA 0 GBP
ACTIAS21228G25 ACTIAS21228G5
1-(4-Cyanophenyl)piperazine
1-(4-Cyanophenyl)piperazine
Formula: C₁₁H₁₃N₃
MDL Number: MFCD01320890
CAS Number: 68104-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR