1-(3-Cyanophenyl)piperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Piperazin-1-yl)benzonitrile

AS22617G1 AS22617G5
ACTIAS22617G1EA 68 GBP
ACTIAS22617G1 ACTIAS22617G5
1-(3-Cyanophenyl)piperazine
1-(3-Cyanophenyl)piperazine
Formula: C₁₁H₁₃N₃
MW: 187.2410 g/mol
MDL Number: MFCD06796181
CAS Number: 178928-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR