1-(2-Cyanophenyl)piperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Piperazin-1-yl)benzonitrile, 1-(2-Cyanophenyl)piperazine, 2-(1-Piperazinyl)benzonitrile

AS20287G25 AS20287G5
ACTIAS20287G25EA 285 GBP
ACTIAS20287G25 ACTIAS20287G5
1-(2-Cyanophenyl)piperazine
1-(2-Cyanophenyl)piperazine
Formula: C₁₁H₁₃N₃
MW: 187.25 g/mol
MDL Number: MFCD00040793
CAS Number: 111373-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR